Assay Details
| Gene Symbol: PRELID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008917.4 | XP_004008966.2 | 4 - 5 | 568 | 72 | |
| XM_027970359.1 | XP_027826160.1 | 4 - 5 | 380 | 72 | ||
More Information
Set Membership: |
|

PRELID2
Genomic Map