Assay Details
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003312.4 | XP_004003361.1 | 4 - 5 | 742 | 109 | |
| XM_004003313.4 | XP_004003362.2 | 4 - 5 | 612 | 109 | ||
More Information
Set Membership: |
|

CLDN18
Genomic Map