Assay Details
Gene Symbol: RBAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961440.1 | XP_027817241.1 | 1 - 2 | 1097 | 116 | |
XM_027961441.1 | XP_027817242.1 | 3 - 4 | 638 | 116 |

More Information
Set Membership: |
![]() ![]() |
