Assay Details
| Gene Symbol: ZNF613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015404.4 | XP_004015453.4 | 6 | 2988 | 104 | |
| XM_027977468.1 | XP_027833269.1 | 5 | 3433 | 104 | ||
| XM_027977470.1 | XP_027833271.1 | 6 | 2916 | 104 | ||
| XM_027977471.1 | XP_027833272.1 | 5 | 2870 | 104 | ||
| XM_027977472.1 | XP_027833273.1 | 6 | 2173 | 104 | ||
| XM_027977473.1 | XP_027833274.1 | 5 | 2192 | 104 | ||
| XM_027977474.1 | XP_027833275.1 | 5 | 2047 | 104 | ||
| XM_027977475.1 | XP_027833276.1 | 5 | 3449 | 104 | ||
More Information
Set Membership: |
|

ZNF613
Genomic Map