Assay Details
| Gene Symbol: AHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965439.1 | XP_027821240.1 | 7 | 6153 | 166 | |
| XM_027965440.1 | XP_027821241.1 | 6 | 5822 | 166 | ||
| XM_027965441.1 | XP_027821242.1 | 7 | 7640 | 166 | ||
More Information
Set Membership: |
|

AHDC1
Genomic Map