Assay Details
| Gene Symbol: SET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966351.1 | XP_027822152.1 | 8 | 2521 | 101 | |
| XM_027966352.1 | XP_027822153.1 | 8 | 2473 | 101 | ||
More Information
Set Membership: |
|

SET
Genomic Map