Assay Details
| Gene Symbol: PCGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270439.2 | XP_008268661.1 | 9 - 10 | 1092 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCGF6| Gene Symbol: PCGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270439.2 | XP_008268661.1 | 9 - 10 | 1092 | 64 | |
Set Membership: |
|