Assay Details
Gene Symbol: UPF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720125.3 | XP_002720171.1 | 5 - 6 | 745 | 94 | |
XM_002720126.3 | XP_002720172.1 | 5 - 6 | 730 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
