Assay Details
| Gene Symbol: EGR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249866.2 | XP_008248088.2 | 337 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008249866.2 transcripts.
Set Membership: |
|

EGR3
Genomic Map