Assay Details
| Gene Symbol: ZDHHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260501.2 | XP_008258723.1 | 4 - 5 | 805 | 66 | |
| XM_008260502.2 | XP_008258724.1 | 4 - 5 | 804 | 66 | ||
| XM_017343760.1 | XP_017199249.1 | 4 - 5 | 684 | 66 | ||
More Information
Set Membership: |
|

ZDHHC3
Genomic Map