Assay Details
| Gene Symbol: PDGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195762.1 | NP_001182691.1 | 5 - 6 | 775 | 80 | |
| XM_002708534.3 | XP_002708580.1 | 5 - 6 | 1185 | 80 | ||
| GenBank mRNA | AY347260.1 | - | 4 - 5 | 607 | 80 | |
More Information
Set Membership: |
|

PDGFD
Genomic Map