Assay Details
| Gene Symbol: LBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195719.1 | NP_001182648.1 | 7 - 8 | 754 | 62 | |
| GenBank mRNA | M35534.1 | - | 7 - 8 | 754 | 62 | |
More Information
Set Membership: |
|

LBP
Genomic Map