Assay Details
| Gene Symbol: FTH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274404.2 | XP_008272626.1 | 352 | 77 | ||
| GenBank mRNA | M63912.1 | - | 1 - 2 | 53 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FTH1| Gene Symbol: FTH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274404.2 | XP_008272626.1 | 352 | 77 | ||
| GenBank mRNA | M63912.1 | - | 1 - 2 | 53 | 77 | |
Set Membership: |
|