Assay Details
| Gene Symbol: MMP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256066.2 | XP_008254288.1 | 4 - 5 | 880 | 90 | |
| GenBank mRNA | EF472906.1 | - | 3 - 4 | 533 | 90 | |
More Information
Set Membership: |
|

MMP24
Genomic Map