Assay Details
| Gene Symbol: CALB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082112.1 | NP_001075581.1 | 4 - 5 | 493 | 73 | |
| GenBank mRNA | AF391152.1 | - | 4 - 5 | 493 | 73 | |
More Information
Set Membership: |
|

CALB1
Genomic Map