Assay Details
| Gene Symbol: KCNJ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082140.1 | NP_001075609.1 | 1 - 2 | 376 | 92 | |
| XM_008259337.2 | XP_008257559.1 | 2 - 3 | 536 | 92 | ||
| GenBank mRNA | AF417509.1 | - | 1 - 2 | 377 | 92 | |
| AJ309931.1 | - | 1 - 2 | 60 | 92 | ||
| AY072063.1 | - | 1 - 2 | 376 | 92 | ||
More Information
Set Membership: |
|

KCNJ8
Genomic Map