Assay Details
| Gene Symbol: KCNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082180.1 | NP_001075649.1 | 2 - 3 | 756 | 96 | |
| GenBank mRNA | AY752982.1 | - | 2 - 3 | 756 | 96 | |
More Information
Set Membership: |
|

KCNK1
Genomic Map