Assay Details
| Gene Symbol: DCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082330.1 | NP_001075799.1 | 2 - 3 | 340 | 118 | |
| XM_008256721.1 | XP_008254943.1 | 3 - 4 | 534 | 118 | ||
| XM_008256722.2 | XP_008254944.1 | 3 - 4 | 528 | 118 | ||
| GenBank mRNA | S76584.1 | - | 2 - 3 | 340 | 118 | |
| U03394.1 | - | 2 - 3 | 214 | 118 | ||
More Information
Set Membership: |
|

DCN
Genomic Map