Assay Details
| Gene Symbol: CYP8B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082622.1 | NP_001076091.1 | 1 | 1550 | 76 | |
| GenBank mRNA | Y08269.1 | - | 1550 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP8B1| Gene Symbol: CYP8B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082622.1 | NP_001076091.1 | 1 | 1550 | 76 | |
| GenBank mRNA | Y08269.1 | - | 1550 | 76 | ||
Set Membership: |
|