Assay Details
| Gene Symbol: TRPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082779.1 | NP_001076248.1 | 2 - 3 | 1157 | 60 | |
| GenBank mRNA | AJ006203.1 | - | 2 - 3 | 1157 | 60 | |
More Information
Set Membership: |
|

TRPC5
Genomic Map