Assay Details
| Gene Symbol: WNT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082665.1 | NP_001076134.1 | 3 - 4 | 686 | 70 | |
| GenBank mRNA | DQ388600.1 | - | 3 - 4 | 686 | 70 | |
More Information
Set Membership: |
|

WNT5A
Genomic Map