Assay Details
| Gene Symbol: TNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109818.1 | NP_001103288.1 | 6 - 7 | 739 | 91 | |
| GenBank mRNA | EF608503.1 | - | 6 - 7 | 739 | 91 | |
More Information
Set Membership: |
|

TNMD
Genomic Map