Assay Details
| Gene Symbol: SYPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081986.1 | NP_001075455.1 | 6 | 2854 | 105 | |
| GenBank mRNA | AB004816.1 | - | 6 | 2854 | 105 | |
More Information
Set Membership: |
|

SYPL2
Genomic Map