Assay Details
| Gene Symbol: GCSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082720.1 | NP_001076189.1 | 1 | 450 | 105 | |
| GenBank mRNA | AF231451.1 | - | 1 | 450 | 105 | |
| Gene Symbol: GCSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342388.1 | XP_017197877.1 | 4 | 321 | 105 | |
More Information
Set Membership: |
|

GCSH
Genomic Map