Assay Details
| Gene Symbol: TLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082781.1 | NP_001076250.1 | 2 | 2949 | 110 | |
| GenBank mRNA | AY101393.1 | - | 2 | 2949 | 110 | |
More Information
Set Membership: |
|

TLR2
Genomic Map