Assay Details
| Gene Symbol: CRHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163115.2 | NP_001156587.1 | 4 - 5 | 347 | 61 | |
| GenBank mRNA | EU519319.1 | - | 4 - 5 | 349 | 61 | |
More Information
Set Membership: |
|

CRHR1
Genomic Map