Assay Details
| Gene Symbol: EGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347349.1 | XP_017202838.1 | 19 - 20 | 3354 | 80 | |
| GenBank mRNA | AF333181.1 | - | 1 - 2 | 59 | 80 | |
More Information
Set Membership: |
|

EGF
Genomic Map