Assay Details
| Gene Symbol: APOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160281.2 | NP_001153753.1 | 251 | 98 | ||
| GenBank mRNA | EU673468.1 | - | 254 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APOC1| Gene Symbol: APOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160281.2 | NP_001153753.1 | 251 | 98 | ||
| GenBank mRNA | EU673468.1 | - | 254 | 98 | ||
Set Membership: |
|