Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171275.1 | NP_001164746.1 | 2 - 3 | 1022 | 79 | |
| XM_008261491.2 | XP_008259713.1 | 2 - 3 | 1050 | 79 | ||
More Information
Set Membership: |
|

STEAP2
Genomic Map