Assay Details
| Gene Symbol: SERPINB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171286.1 | NP_001164757.1 | 6 - 7 | 741 | 73 | |
| XM_008261387.2 | XP_008259609.1 | 7 - 8 | 809 | 73 | ||
| XM_008261388.2 | XP_008259610.1 | 7 - 8 | 848 | 73 | ||
| XM_017343654.1 | XP_017199143.1 | 7 - 8 | 886 | 73 | ||
More Information
Set Membership: |
|

SERPINB7
Genomic Map