Assay Details
| Gene Symbol: GABRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319574.1 | NP_001306503.1 | 6 - 7 | 710 | 75 | |
| XM_002710356.3 | XP_002710402.2 | 7 - 8 | 1193 | 75 | ||
| GenBank mRNA | KP657519.1 | - | 6 - 7 | 710 | 75 | |
| KP657520.1 | - | 6 - 7 | 710 | 75 | ||
| KP657521.1 | - | 3 - 4 | 287 | 75 | ||
| KP657522.1 | - | 3 - 4 | 449 | 75 | ||
More Information
Set Membership: |
|

GABRA1
Genomic Map