Assay Details
| Gene Symbol: LOC100356068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329075.1 | NP_001316004.1 | 2 - 3 | 288 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100356068| Gene Symbol: LOC100356068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329075.1 | NP_001316004.1 | 2 - 3 | 288 | 107 | |
Set Membership: |
|