Assay Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711921.3 | XP_002711967.1 | 7 | 3673 | 73 | |
| XM_008258006.2 | XP_008256228.1 | 7 | 3677 | 73 | ||
| XM_008258008.2 | XP_008256230.1 | 7 | 3533 | 73 | ||
| XM_017342585.1 | XP_017198074.1 | 6 | 3967 | 73 | ||
More Information
Set Membership: |
|

NYAP1
Genomic Map