Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723078.3 | XP_002723124.1 | 1072 | 61 | ||
| XM_017339079.1 | XP_017194568.1 | 1075 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCZ1| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723078.3 | XP_002723124.1 | 1072 | 61 | ||
| XM_017339079.1 | XP_017194568.1 | 1075 | 61 | |||
Set Membership: |
|