Assay Details
| Gene Symbol: RAB3IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256779.2 | XP_008255001.1 | 7 - 8 | 1251 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB3IP| Gene Symbol: RAB3IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256779.2 | XP_008255001.1 | 7 - 8 | 1251 | 70 | |
Set Membership: |
|