Assay Details
Gene Symbol: TMEM263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711423.3 | XP_002711469.1 | 1 - 2 | 341 | 119 | |
XM_008257103.2 | XP_008255325.1 | 2 - 3 | 357 | 119 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002711423.3, XM_008257103.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
