Assay Details
| Gene Symbol: NDUFAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709761.3 | XP_002709807.1 | 2 - 3 | 275 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFAF7| Gene Symbol: NDUFAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709761.3 | XP_002709807.1 | 2 - 3 | 275 | 62 | |
Set Membership: |
|