Assay Details
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273926.2 | XP_008272148.2 | 660 | 86 | ||
| XM_017350386.1 | XP_017205875.1 | 660 | 86 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SFRP1| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008273926.2 | XP_008272148.2 | 660 | 86 | ||
| XM_017350386.1 | XP_017205875.1 | 660 | 86 | |||
Set Membership: |
|