Assay Details
| Gene Symbol: SREK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262268.2 | XP_008260490.1 | 2 - 3 | 261 | 71 | |
| XM_017344748.1 | XP_017200237.1 | 2 - 3 | 99 | 71 | ||
More Information
Set Membership: |
|

SREK1IP1
Genomic Map