Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261537.2 | XP_008259759.1 | 3 - 4 | 485 | 64 | |
| XM_008261538.2 | XP_008259760.1 | 3 - 4 | 485 | 64 | ||
More Information
Set Membership: |
|

FAM221A
Genomic Map