Assay Details
| Gene Symbol: PCDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712974.3 | XP_002713020.1 | 1 - 2 | 3873 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDH8| Gene Symbol: PCDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712974.3 | XP_002713020.1 | 1 - 2 | 3873 | 93 | |
Set Membership: |
|