Assay Details
| Gene Symbol: PCDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712974.3 | XP_002713020.1 | 2 - 3 | 4086 | 94 | |
| XM_002712975.3 | XP_002713021.1 | 2 - 3 | 3794 | 94 | ||
More Information
Set Membership: |
|

PCDH8
Genomic Map