Assay Details
| Gene Symbol: TIMMDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716669.3 | XP_002716715.1 | 1 - 2 | 416 | 74 | |
| XM_017347070.1 | XP_017202559.1 | 1 - 2 | 416 | 74 | ||
| XM_017347071.1 | XP_017202560.1 | 1 - 2 | 416 | 74 | ||
More Information
Set Membership: |
|

TIMMDC1
Genomic Map