Assay Details
| Gene Symbol: TMEM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709672.3 | XP_002709718.1 | 5 - 6 | 465 | 57 | |
| XM_017340384.1 | XP_017195873.1 | 4 - 5 | 375 | 57 | ||
More Information
Set Membership: |
|

TMEM150A
Genomic Map