Assay Details
| Gene Symbol: GPR63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263290.2 | XP_008261512.2 | 1 - 2 | 223 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008263290.2 transcripts.
Set Membership: |
|

GPR63
Genomic Map