Assay Details
| Gene Symbol: ANKRD34B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344517.1 | XP_017200006.1 | 1 - 2 | 111 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017344517.1 transcripts.
Set Membership: |
|

ANKRD34B
Genomic Map