Assay Details
Gene Symbol: FAM71D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008271843.2 | XP_008270065.1 | 4 - 5 | 626 | 76 | |
XM_008271844.2 | XP_008270066.1 | 4 - 5 | 626 | 76 | ||
XM_008271847.2 | XP_008270069.1 | 4 - 5 | 625 | 76 | ||
XM_008271848.2 | XP_008270070.1 | 4 - 5 | 565 | 76 | ||
XM_008271850.2 | XP_008270072.1 | 2 - 3 | 384 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
