Assay Details
| Gene Symbol: TMEM95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270788.2 | XP_008269010.1 | 5 - 6 | 504 | 82 | |
| XM_017348876.1 | XP_017204365.1 | 4 - 5 | 591 | 82 | ||
| XM_017348877.1 | XP_017204366.1 | 6 - 7 | 585 | 82 | ||
More Information
Set Membership: |
|

TMEM95
Genomic Map