Assay Details
| Gene Symbol: LOC100338764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717341.3 | XP_002717387.1 | 12 - 13 | 3019 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100338764| Gene Symbol: LOC100338764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717341.3 | XP_002717387.1 | 12 - 13 | 3019 | 98 | |
Set Membership: |
|