Assay Details
| Gene Symbol: LOC100338764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717341.3 | XP_002717387.1 | 3 - 4 | 1108 | 101 | |
| XM_008268232.2 | XP_008266454.1 | 3 - 4 | 1108 | 101 | ||
More Information
Set Membership: |
|

LOC100338764
Genomic Map